Product Details

SNP ID
rs140315337
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:85250607 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATATTTAGCCTCTTCCCTAAGTCC[A/T]GTCTTCCTCCTCATACAACATACTC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
C1orf52 PubMed Links

Gene Details

Gene
C1orf52
Gene Name
chromosome 1 open reading frame 52
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198077.3 2610 UTR 3 NP_932343.1

View Full Product Details