Product Details

SNP ID
rs140410603
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:23193437 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCAGCCGAGTGCGAGTGCCCCTC[A/C]TGGAGGCTGGAGTTGAGCGAGCAGA
Phenotype
MIM: 182133
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HTR1D PubMed Links

Gene Details

Gene
HTR1D
Gene Name
5-hydroxytryptamine receptor 1D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000864.4 1293 Silent Mutation NP_000855.1

View Full Product Details