Product Details

SNP ID
rs140739406
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:205718475 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCTTTGCCTTTCACTGGACTTGG[C/T]GTCACTGAAAATAGAAAAATAATTT
Phenotype
MIM: 611912
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NUCKS1 PubMed Links

Gene Details

Gene
NUCKS1
Gene Name
nuclear casein kinase and cyclin dependent kinase substrate 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022731.4 781 Silent Mutation ACA,ACG T179T NP_073568.2
XM_005245453.1 781 Silent Mutation ACA,ACG T178T XP_005245510.1

View Full Product Details