Product Details

SNP ID
rs140768884
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:167723011 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCTCCTTTTTCTTACCAAATATT[A/G]TGACTCAAGCTTAAGGTAAACCTCG
Phenotype
MIM: 604376
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MPZL1 PubMed Links

Gene Details

Gene
MPZL1
Gene Name
myelin protein zero like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146191.1 Intron NP_001139663.1
NM_003953.5 Intron NP_003944.1
NM_024569.4 Intron NP_078845.3

View Full Product Details