Product Details

SNP ID
rs140771260
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:151400525 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCTGACCAGGGCCATGTACAGCC[A/G]GCGCTCGAAGATGAACCCTTTGTGG
Phenotype
MIM: 614787 MIM: 602177
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
POGZ PubMed Links

Gene Details

Gene
POGZ
Gene Name
pogo transposable element with ZNF domain
There are no transcripts associated with this gene.

Gene
PSMB4
Gene Name
proteasome subunit beta 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002796.2 454 Missense Mutation CAG,CGG Q144R NP_002787.2

View Full Product Details