Product Details

SNP ID
rs140904774
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:6525880 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTTCATTTTTCGACAGAACTTAG[A/G]TCTTCGGTATGGTTTCTCTTTATGT
Phenotype
MIM: 611101
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NOL9 PubMed Links

Gene Details

Gene
NOL9
Gene Name
nucleolar protein 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024654.4 1105 Missense Mutation CCT,TCT P695S NP_078930.3
XM_005263493.3 1105 Missense Mutation CCT,TCT P408S XP_005263550.1
XM_011542147.2 1105 Missense Mutation CCT,TCT P325S XP_011540449.1
XM_017002337.1 1105 Intron XP_016857826.1
XM_017002338.1 1105 Missense Mutation CCT,TCT P325S XP_016857827.1
XM_017002339.1 1105 Missense Mutation CCT,TCT P325S XP_016857828.1
Gene
PLEKHG5
Gene Name
pleckstrin homology and RhoGEF domain containing G5
There are no transcripts associated with this gene.

View Full Product Details