Product Details

SNP ID
rs140949805
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:108929928 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTTAAAAATTCAGGGAAAAAATC[A/G]GCAGACCATTAGTTACTATGGATTT
Phenotype
MIM: 609245
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLCC1 PubMed Links

Gene Details

Gene
CLCC1
Gene Name
chloride channel CLIC like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001048210.2 2539 UTR 3 NP_001041675.1
NM_001278202.1 2539 UTR 3 NP_001265131.1
NM_001278203.1 2539 UTR 3 NP_001265132.1
NM_015127.4 2539 UTR 3 NP_055942.1
Gene
GPSM2
Gene Name
G-protein signaling modulator 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321038.1 2539 Silent Mutation TCA,TCG S681S NP_001307967.1
NM_001321039.1 2539 Silent Mutation TCA,TCG S681S NP_001307968.1
NM_013296.4 2539 Silent Mutation TCA,TCG S681S NP_037428.3
XM_006710589.2 2539 Silent Mutation TCA,TCG S662S XP_006710652.1
XM_011541301.2 2539 Silent Mutation TCA,TCG S681S XP_011539603.1
XM_011541302.2 2539 Silent Mutation TCA,TCG S681S XP_011539604.1
XM_011541303.2 2539 Intron XP_011539605.1
XM_017001097.1 2539 Silent Mutation TCA,TCG S681S XP_016856586.1
XM_017001098.1 2539 Silent Mutation TCA,TCG S681S XP_016856587.1

View Full Product Details