Product Details

SNP ID
rs141056335
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:53218456 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACACAGCTTGCACTTCTGGACCA[C/T]GGAAGCACTGCCACGGCCCCCCTTC
Phenotype
MIM: 600650 MIM: 602603
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C1orf123 PubMed Links

Gene Details

Gene
C1orf123
Gene Name
chromosome 1 open reading frame 123
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304759.1 194 Missense Mutation NP_001291688.1
NM_001304760.1 194 Missense Mutation NP_001291689.1
NM_017887.2 194 Missense Mutation NP_060357.1
Gene
CPT2
Gene Name
carnitine palmitoyltransferase 2
There are no transcripts associated with this gene.

Gene
MAGOH
Gene Name
mago homolog, exon junction complex core component
There are no transcripts associated with this gene.

View Full Product Details