Product Details

SNP ID
rs141209616
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:10433899 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTCTTTGCGAGGAAGTTGCATTG[A/G]ACAAAGAGATGCAGTTCAGCAAACA
Phenotype
MIM: 609130
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
APITD1 PubMed Links

Gene Details

Gene
APITD1
Gene Name
apoptosis-inducing, TAF9-like domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_199294.2 525 Missense Mutation AAC,GAC N37D NP_954988.1
Gene
APITD1-CORT
Gene Name
APITD1-CORT readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243768.1 525 Intron NP_001230697.1
NM_001270517.1 525 Missense Mutation AAC,GAC N37D NP_001257446.1
NM_198544.3 525 Missense Mutation AAC,GAC N37D NP_940946.1
NM_199006.2 525 Missense Mutation AAC,GAC N37D NP_950171.2

View Full Product Details