Product Details

SNP ID
rs141403253
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:54999288 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGCCATGATCGGCCCCAGGTCTA[C/T]GGCACCTTCTATGCCATGGGCAGCG
Phenotype
MIM: 606412
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BSND PubMed Links

Gene Details

Gene
BSND
Gene Name
barttin CLCNK type accessory beta subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_057176.2 345 Silent Mutation TAC,TAT Y34Y NP_476517.1
Gene
TMEM61
Gene Name
transmembrane protein 61
There are no transcripts associated with this gene.

View Full Product Details