Product Details

SNP ID
rs141403958
Assay Type
Functionally tested
NCBI dbSNP Submissions
18
Location
Chr.1:149886630 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTAGGGGCCGGAGCGGATTTTGCC[G/T]GTTCAGGCATGGTAAGACACAGTAC
Phenotype
MIM: 615014 MIM: 602797 MIM: 601831
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HIST2H2AB PubMed Links

Gene Details

Gene
HIST2H2AB
Gene Name
histone cluster 2, H2ab
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175065.2 53 Intron NP_778235.1
Gene
HIST2H2AC
Gene Name
histone cluster 2, H2ac
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003517.2 53 Intron NP_003508.1
Gene
HIST2H2BE
Gene Name
histone cluster 2, H2be
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003528.2 53 Missense Mutation CAG,CCG Q4P NP_003519.1

View Full Product Details