Product Details
- SNP ID
-
rs141424928
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
6
- Location
-
Chr.1:10008245 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGAAAAGAAGAACAGAGGCTGGACC[C/T]ATTGGATCGAAGGAGACAAACTCCA
- Phenotype
-
MIM: 608604
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RBP7
PubMed Links
Gene Details
- Gene
- RBP7
- Gene Name
- retinol binding protein 7
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_052960.2 |
387 |
Missense Mutation |
CAT,TAT |
H109Y |
NP_443192.1 |
View Full Product Details