Product Details

SNP ID
rs141438850
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:27356389 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCTCCAGCCAAGGCCCACTCACT[C/T]GGAGGCTCTGGGGCCAGGACATAGG
Phenotype
MIM: 604468 MIM: 608042
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAP3K6 PubMed Links

Gene Details

Gene
MAP3K6
Gene Name
mitogen-activated protein kinase kinase kinase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297609.1 3881 Silent Mutation CCA,CCG P1204P NP_001284538.1
NM_004672.4 3881 Silent Mutation CCA,CCG P1212P NP_004663.3
XM_017002771.1 3881 Silent Mutation CCA,CCG P793P XP_016858260.1
XM_017002772.1 3881 Intron XP_016858261.1
Gene
SYTL1
Gene Name
synaptotagmin like 1
There are no transcripts associated with this gene.

View Full Product Details