Product Details

SNP ID
rs141499812
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:18635119 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCTCCACCTCTGAAGCAGGTGGC[A/G]ACTCCGGATGTAGAGAAAAAGATTG
Phenotype
MIM: 167410
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PAX7 PubMed Links

Gene Details

Gene
PAX7
Gene Name
paired box 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135254.1 928 Silent Mutation GCA,GCG A110A NP_001128726.1
NM_002584.2 928 Silent Mutation GCA,GCG A110A NP_002575.1
NM_013945.2 928 Silent Mutation GCA,GCG A110A NP_039236.1

View Full Product Details