Product Details

SNP ID
rs141501072
Assay Type
Functionally Tested
NCBI dbSNP Submissions
4
Location
Chr.1:173481344 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTCAGATGGGGCATTCTCTTCTC[A/C]CACCCTCGGGACTTTACCCCAGTGT
Phenotype
MIM: 602316
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
LOC100506023 PubMed Links

Gene Details

Gene
LOC100506023
Gene Name
uncharacterized LOC100506023
There are no transcripts associated with this gene.

Gene
PRDX6
Gene Name
peroxiredoxin 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004905.2 Intron NP_004896.1

View Full Product Details