Product Details

SNP ID
rs141567577
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:145867357 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCCTCTTCCATGCTAAGGATTTG[G/C]AGCAGGATATTGTACACTTCATGTT
Phenotype
MIM: 605987
Polymorphism
G/C, Transversion substitution
Allele Nomenclature
Literature Links
ANKRD35 PubMed Links

Gene Details

Gene
ANKRD35
Gene Name
ankyrin repeat domain 35
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001280799.1 3087 Silent Mutation CTC,CTG L903L NP_001267728.1
NM_144698.4 3087 Silent Mutation CTC,CTG L993L NP_653299.4
Gene
PIAS3
Gene Name
protein inhibitor of activated STAT 3
There are no transcripts associated with this gene.

View Full Product Details