Product Details

SNP ID
rs141731322
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:227921698 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGGCACCTACGGCCAGCGGTGCC[C/T]GGGGAGAAGCGGCCAGCCAGGCAGA
Phenotype
MIM: 602863
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
WNT9A PubMed Links

Gene Details

Gene
WNT9A
Gene Name
Wnt family member 9A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003395.2 1489 Silent Mutation NP_003386.1
XM_011544271.2 1489 Silent Mutation XP_011542573.1

View Full Product Details