Product Details

SNP ID
rs141833562
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:231338468 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGCGCGATCATGCCCAGGAGTCC[C/T]TGTCGCTAGTGGACGCGTCGTGGGA
Phenotype
MIM: 615283 MIM: 616086
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EXOC8 PubMed Links

Gene Details

Gene
EXOC8
Gene Name
exocyst complex component 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175876.4 533 Intron NP_787072.2
Gene
SPRTN
Gene Name
SprT-like N-terminal domain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010984.3 533 Silent Mutation CTG,TTG L29L NP_001010984.1
NM_001261462.2 533 Silent Mutation CTG,TTG L29L NP_001248391.1
NM_032018.6 533 Silent Mutation CTG,TTG L29L NP_114407.3
XM_006711818.2 533 Silent Mutation CTG,TTG L29L XP_006711881.1
XM_011544289.2 533 Intron XP_011542591.1

View Full Product Details