Product Details

SNP ID
rs141882799
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:35825684 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAAACTCTTATTTCTTCAGGTACA[C/T]CCCAGTGGGCCGTTCCTTTTTCTCA
Phenotype
MIM: 607356
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AGO4 PubMed Links

Gene Details

Gene
AGO4
Gene Name
argonaute 4, RISC catalytic component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017629.3 569 Missense Mutation ACC,ATC T165I NP_060099.2
XM_005270578.3 569 Missense Mutation ACC,ATC T98I XP_005270635.1
XM_005270579.3 569 Missense Mutation ACC,ATC T98I XP_005270636.1
XM_011540883.2 569 Missense Mutation ACC,ATC T15I XP_011539185.1
XM_011540884.2 569 Intron XP_011539186.1
XM_017000529.1 569 Missense Mutation ACC,ATC T165I XP_016856018.1

View Full Product Details