Product Details

SNP ID
rs141952418
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:247841140 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACACATACATGGAGATCATGGTCC[C/T]GTAGTAGAGAGTGACAACAGCCAGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR11L1 PubMed Links

Gene Details

Gene
OR11L1
Gene Name
olfactory receptor family 11 subfamily L member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001959.1 757 Missense Mutation AGG,GGG R253G NP_001001959.1

View Full Product Details