Product Details

SNP ID
rs141969937
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:38045741 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCTTCTTGCGCTTGCGGCCCTGCG[C/T]CGCGATCTTGTCCAGGTTGGTGGGG
Phenotype
MIM: 602479
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC107985103 PubMed Links

Gene Details

Gene
LOC107985103
Gene Name
translation initiation factor IF-2-like
There are no transcripts associated with this gene.

Gene
POU3F1
Gene Name
POU class 3 homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002699.3 1038 Missense Mutation ACG,GCG T335A NP_002690.3

View Full Product Details