Product Details

SNP ID
rs142007100
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:160218325 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTCATTTCCAACCCTAGCTTACC[C/T]GGTGATGGCGTCTCTGTCTCAGGTG
Phenotype
MIM: 615820 MIM: 603434
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DCAF8 PubMed Links

Gene Details

Gene
DCAF8
Gene Name
DDB1 and CUL4 associated factor 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015726.3 1888 Missense Mutation CAG,CGG Q559R NP_056541.2
Gene
LOC729867
Gene Name
uncharacterized LOC729867
There are no transcripts associated with this gene.

Gene
PEA15
Gene Name
phosphoprotein enriched in astrocytes 15
There are no transcripts associated with this gene.

View Full Product Details