Product Details

SNP ID
rs142146082
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:231335739 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTCTGTCTGATAAAAGCTTTCTT[A/C]TCTGGATCCTGCTCACATCGAAGAC
Phenotype
MIM: 615283 MIM: 616086
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
EXOC8 PubMed Links

Gene Details

Gene
EXOC8
Gene Name
exocyst complex component 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175876.4 2134 Silent Mutation NP_787072.2
Gene
SPRTN
Gene Name
SprT-like N-terminal domain
There are no transcripts associated with this gene.

View Full Product Details