Product Details

SNP ID
rs142156708
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:108753104 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTTACCACTAAGCTTTTGGCATC[A/G]TGTTGCAAAATGACAGATCTTCTAG
Phenotype
MIM: 608339
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
STXBP3 PubMed Links

Gene Details

Gene
STXBP3
Gene Name
syntaxin binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007269.2 216 Silent Mutation TCA,TCG S47S NP_009200.2

View Full Product Details