Product Details

SNP ID
rs142161528
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:43283064 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCGAGCCCCAGTGCCTGGGCCAGG[A/G]GCCACAGCAGACGCTGTGGGGAGCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1orf210 PubMed Links

Gene Details

Gene
C1orf210
Gene Name
chromosome 1 open reading frame 210
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164829.1 226 Silent Mutation GCC,GCT A21A NP_001158301.1
NM_182517.2 226 Silent Mutation GCC,GCT A21A NP_872323.1
XM_011540802.1 226 Silent Mutation GCC,GCT A21A XP_011539104.1
Gene
TMEM125
Gene Name
transmembrane protein 125
There are no transcripts associated with this gene.

View Full Product Details