Product Details

SNP ID
rs142216400
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:100086334 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGACTTTTTTTCTATTTTATTTTT[G/T]TCTTTAAAGTTTGATACCCGGTGCC
Phenotype
MIM: 609321
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MFSD14A PubMed Links

Gene Details

Gene
MFSD14A
Gene Name
major facilitator superfamily domain containing 14A
There are no transcripts associated with this gene.

Gene
SASS6
Gene Name
SAS-6 centriolar assembly protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304829.1 Intron NP_001291758.1
NM_194292.2 Intron NP_919268.1
XM_017000486.1 Intron XP_016855975.1

View Full Product Details