Product Details

SNP ID
rs142245176
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:32826243 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTGTCAGAGGGAGAAGAAGATGA[C/T]GGTGATGTAAATTACACAGAGGAAG
Phenotype
MIM: 611889 MIM: 603623
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
S100PBP PubMed Links

Gene Details

Gene
S100PBP
Gene Name
S100P binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256121.1 370 Silent Mutation GAC,GAT D48D NP_001243050.1
NM_022753.3 370 Silent Mutation GAC,GAT D48D NP_073590.2
XM_011541961.2 370 Silent Mutation GAC,GAT D48D XP_011540263.1
XM_011541962.2 370 Silent Mutation GAC,GAT D48D XP_011540264.1
XM_011541963.1 370 Silent Mutation GAC,GAT D48D XP_011540265.1
XM_017002098.1 370 Silent Mutation GAC,GAT D48D XP_016857587.1
Gene
YARS
Gene Name
tyrosyl-tRNA synthetase
There are no transcripts associated with this gene.

View Full Product Details