Product Details

SNP ID
rs142288232
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:31619252 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTCCCCCCTGGCAGCAGAGAGCC[A/G]TCCCCTGTGCCTCCAGACTATGAAG
Phenotype
MIM: 602392
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HCRTR1 PubMed Links

Gene Details

Gene
HCRTR1
Gene Name
hypocretin receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001525.2 936 Silent Mutation CCA,CCG P20P NP_001516.2
XM_017001105.1 936 Silent Mutation CCA,CCG P20P XP_016856594.1
XM_017001106.1 936 Silent Mutation CCA,CCG P20P XP_016856595.1
XM_017001107.1 936 Silent Mutation CCA,CCG P20P XP_016856596.1

View Full Product Details