Product Details

SNP ID
rs142288768
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:155067409 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAGAGTTCTGGCCAGTGCTTGAG[A/G]CTCCAGGTGTCTGTCTGCTGCAAGG
Phenotype
MIM: 605548 MIM: 601380
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADAM15 PubMed Links

Gene Details

Gene
ADAM15
Gene Name
ADAM metallopeptidase domain 15
There are no transcripts associated with this gene.

Gene
EFNA4
Gene Name
ephrin A4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005227.2 525 Silent Mutation AGA,AGG R146R NP_005218.1
NM_182689.1 525 Silent Mutation AGA,AGG R146R NP_872631.1
NM_182690.2 525 Silent Mutation AGA,AGG R146R NP_872632.2
Gene
LOC100505666
Gene Name
uncharacterized LOC100505666
There are no transcripts associated with this gene.

View Full Product Details