Product Details

SNP ID
rs142488044
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:27332097 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACAACATGGCCTTTGGAAAGCCT[A/G]CCAAGTAAGTGATGAACACCCATGT
Phenotype
MIM: 608042
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACTG1P20 PubMed Links

Gene Details

Gene
ACTG1P20
Gene Name
actin gamma 1 pseudogene 20
There are no transcripts associated with this gene.

Gene
SYTL1
Gene Name
synaptotagmin like 1
There are no transcripts associated with this gene.

Gene
TMEM222
Gene Name
transmembrane protein 222
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032125.2 360 Missense Mutation ACC,GCC T103A NP_115501.2

View Full Product Details