Product Details
- SNP ID
-
rs142551283
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
7
- Location
-
Chr.1:1460881 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCGCGCCTGGTGAGAATGTATCTTA[A/T]CGAGTATGTTCTTAAGCCGGCCACA
- Phenotype
-
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ATAD3C
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs72896283] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ATAD3C
- Gene Name
- ATPase family, AAA domain containing 3C
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001039211.2 |
1939 |
Missense Mutation |
AAC,ATC |
N315I |
NP_001034300.2 |
View Full Product Details