Product Details

SNP ID
rs142609327
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:109604100 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAACAGATGCAAAGACTCATGCATA[C/T]GATTCTAGTAAGAGGAAACACCATT
Phenotype
MIM: 139370 MIM: 139340 MIM: 611189
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GNAI3 PubMed Links

Gene Details

Gene
GNAI3
Gene Name
G protein subunit alpha i3
There are no transcripts associated with this gene.

Gene
GNAT2
Gene Name
G protein subunit alpha transducin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005272.3 938 Missense Mutation CAT,CGT H242R NP_005263.1
XM_011541264.2 938 Missense Mutation CAT,CGT H242R XP_011539566.1
Gene
MIR197
Gene Name
microRNA 197
There are no transcripts associated with this gene.

View Full Product Details