Product Details
- SNP ID
-
rs142929868
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
8
- Location
-
Chr.1:196951021 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTGGTCCACTCCTCCCAAATGCAG[G/T]TCCACTAGTAAGTGCAATGTTGTTC
- Phenotype
-
MIM: 600889
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CFHR2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs4085749] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CFHR2
- Gene Name
- complement factor H related 2
View Full Product Details