Product Details

SNP ID
rs142946157
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:150983128 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCATGTCTGTGACTGGCGGGAAGA[C/T]GGCACCGTCCCTCACCCAGGAGATC
Phenotype
MIM: 603319 MIM: 606920
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANXA9 PubMed Links

Gene Details

Gene
ANXA9
Gene Name
annexin A9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003568.2 212 Missense Mutation ACG,ATG T8M NP_003559.2
XM_011510058.2 212 Missense Mutation ACG,ATG T8M XP_011508360.1
Gene
CERS2
Gene Name
ceramide synthase 2
There are no transcripts associated with this gene.

View Full Product Details