Product Details

SNP ID
rs143162147
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:203483089 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATCTTCCCTAGAGCTCTAGTTCT[G/T]CCCAACTCTGGCTTCAAAAACATGA
Phenotype
MIM: 601914
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PRELP PubMed Links

Gene Details

Gene
PRELP
Gene Name
proline and arginine rich end leucine rich repeat protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002725.3 Intron NP_002716.1
NM_201348.1 Intron NP_958505.1

View Full Product Details