Product Details
- SNP ID
-
rs143223249
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
10
- Location
-
Chr.1:92246538 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTAGGAAATGGAACTTTCACTTTA[C/T]CCCAATATTTTCTTCAGAGGTAGAC
- Phenotype
-
MIM: 601749
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
C1orf146
PubMed Links
Gene Details
- Gene
- C1orf146
- Gene Name
- chromosome 1 open reading frame 146
- Gene
- GLMN
- Gene Name
- glomulin, FKBP associated protein
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001319683.1 |
1778 |
Missense Mutation |
ATA,GTA |
I579V |
NP_001306612.1 |
NM_053274.2 |
1778 |
Missense Mutation |
ATA,GTA |
I593V |
NP_444504.1 |
XM_005270401.3 |
1778 |
Missense Mutation |
ATA,GTA |
I551V |
XP_005270458.1 |
XM_006710309.2 |
1778 |
Missense Mutation |
ATA,GTA |
I426V |
XP_006710372.1 |
XM_011540546.2 |
1778 |
Missense Mutation |
ATA,GTA |
I593V |
XP_011538848.1 |
XM_017000137.1 |
1778 |
Missense Mutation |
ATA,GTA |
I626V |
XP_016855626.1 |
XM_017000138.1 |
1778 |
Missense Mutation |
ATA,GTA |
I612V |
XP_016855627.1 |
XM_017000139.1 |
1778 |
Missense Mutation |
ATA,GTA |
I591V |
XP_016855628.1 |
XM_017000140.1 |
1778 |
Missense Mutation |
ATA,GTA |
I584V |
XP_016855629.1 |
XM_017000141.1 |
1778 |
Missense Mutation |
ATA,GTA |
I558V |
XP_016855630.1 |
XM_017000142.1 |
1778 |
Missense Mutation |
ATA,GTA |
I412V |
XP_016855631.1 |
XM_017000143.1 |
1778 |
Missense Mutation |
ATA,GTA |
I412V |
XP_016855632.1 |
XM_017000144.1 |
1778 |
Missense Mutation |
ATA,GTA |
I336V |
XP_016855633.1 |
View Full Product Details