Product Details

SNP ID
rs143311715
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:68477944 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGTATGAGTAAGTTGGCAAAGGA[A/G]CAAATAGTCCATCTCCAGGATTTTC
Phenotype
MIM: 612002
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DEPDC1 PubMed Links

Gene Details

Gene
DEPDC1
Gene Name
DEP domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114120.2 2363 Missense Mutation GCT,GTT A714V NP_001107592.1
NM_017779.5 2363 Missense Mutation GCT,GTT A430V NP_060249.2

View Full Product Details