Product Details

SNP ID
rs143326302
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:27879706 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCCTGGAGGAGCTGGTCTCTGCC[A/G]CAACTCAGAGCTCCAAGCAGCTGCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
THEMIS2 PubMed Links

Gene Details

Gene
THEMIS2
Gene Name
thymocyte selection associated family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039477.2 327 Intron NP_001034566.1
NM_001105556.2 327 Missense Mutation ACA,GCA T100A NP_001099026.1
NM_001286113.1 327 Missense Mutation ACA,GCA T100A NP_001273042.1
NM_001286115.1 327 Missense Mutation ACA,GCA T100A NP_001273044.1
NM_004848.3 327 Missense Mutation ACA,GCA T100A NP_004839.2
XM_005246041.3 327 Intron XP_005246098.1
XM_006711050.1 327 Missense Mutation ACA,GCA T100A XP_006711113.1
XM_011542445.2 327 Missense Mutation ACA,GCA T53A XP_011540747.1

View Full Product Details