Product Details

SNP ID
rs143348041
Assay Type
Functionally Tested
NCBI dbSNP Submissions
8
Location
Chr.1:160542598 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAAGAACTTATAAGAAAAATATTT[C/G]TCATTTGCAAAGATTTGGCCATTTT
Phenotype
MIM: 604513
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
CD84 PubMed Links

Gene Details

Gene
CD84
Gene Name
CD84 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184879.1 6414 UTR 3 NP_001171808.1
NM_001184881.1 6414 UTR 3 NP_001171810.1
NM_001184882.1 6414 UTR 3 NP_001171811.1
NM_003874.3 6414 UTR 3 NP_003865.1
XM_011510094.2 6414 UTR 3 XP_011508396.1
XM_011510095.2 6414 UTR 3 XP_011508397.1

View Full Product Details