Product Details

SNP ID
rs143358595
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:153204811 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGAGTCCAAATGCCAGCCCAGCT[A/G]TTTAAAGAAGCTGCTGCAACGCTGT
Phenotype
MIM: 611042
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LELP1 PubMed Links

Gene Details

Gene
LELP1
Gene Name
late cornified envelope like proline rich 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010857.2 227 Missense Mutation TAT,TGT Y35C NP_001010857.1
Gene
SPRR2G
Gene Name
small proline rich protein 2G
There are no transcripts associated with this gene.

View Full Product Details