Product Details
- SNP ID
-
rs143428088
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
12
- Location
-
Chr.1:7786735 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGTTTCTTAGCAAACAGTGAGTTT[G/T]TCCAGATTCTCAGTCAGAATGGAGC
- Phenotype
-
MIM: 603427
MIM: 603657
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PER3
PubMed Links
Gene Details
- Gene
- PER3
- Gene Name
- period circadian clock 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001289861.1 |
428 |
Missense Mutation |
GTC,TTC |
V97F |
NP_001276790.1 |
NM_001289862.1 |
428 |
Missense Mutation |
GTC,TTC |
V97F |
NP_001276791.1 |
NM_001289863.1 |
428 |
Missense Mutation |
GTC,TTC |
V97F |
NP_001276792.1 |
NM_001289864.1 |
428 |
UTR 5 |
|
|
NP_001276793.1 |
NM_016831.2 |
428 |
Missense Mutation |
GTC,TTC |
V97F |
NP_058515.1 |
XM_005263524.3 |
428 |
Missense Mutation |
GTC,TTC |
V97F |
XP_005263581.1 |
XM_011542385.2 |
428 |
Missense Mutation |
GTC,TTC |
V97F |
XP_011540687.1 |
XM_011542390.2 |
428 |
Missense Mutation |
GTC,TTC |
V97F |
XP_011540692.1 |
XM_017002723.1 |
428 |
Missense Mutation |
GTC,TTC |
V97F |
XP_016858212.1 |
XM_017002724.1 |
428 |
Missense Mutation |
GTC,TTC |
V97F |
XP_016858213.1 |
XM_017002725.1 |
428 |
Missense Mutation |
GTC,TTC |
V97F |
XP_016858214.1 |
XM_017002726.1 |
428 |
Missense Mutation |
GTC,TTC |
V97F |
XP_016858215.1 |
XM_017002727.1 |
428 |
Missense Mutation |
GTC,TTC |
V41F |
XP_016858216.1 |
XM_017002728.1 |
428 |
Intron |
|
|
XP_016858217.1 |
XM_017002729.1 |
428 |
Intron |
|
|
XP_016858218.1 |
XM_017002730.1 |
428 |
Intron |
|
|
XP_016858219.1 |
XM_017002731.1 |
428 |
Intron |
|
|
XP_016858220.1 |
XM_017002732.1 |
428 |
Intron |
|
|
XP_016858221.1 |
XM_017002733.1 |
428 |
UTR 5 |
|
|
XP_016858222.1 |
XM_017002734.1 |
428 |
UTR 5 |
|
|
XP_016858223.1 |
XM_017002735.1 |
428 |
UTR 5 |
|
|
XP_016858224.1 |
XM_017002736.1 |
428 |
UTR 5 |
|
|
XP_016858225.1 |
XM_017002737.1 |
428 |
Intron |
|
|
XP_016858226.1 |
XM_017002738.1 |
428 |
Intron |
|
|
XP_016858227.1 |
- Gene
- VAMP3
- Gene Name
- vesicle associated membrane protein 3
There are no transcripts associated with this gene.
View Full Product Details