Product Details
- SNP ID
-
rs143477505
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
6
- Location
-
Chr.1:151199055 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTCGGTTTTTCATCCTTTGATCCCG[C/T]GGTCCCTTCCTGTACCTTGTCCTCA
- Phenotype
-
MIM: 603275
MIM: 600607
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PIP5K1A
PubMed Links
Gene Details
- Gene
- PIP5K1A
- Gene Name
- phosphatidylinositol-4-phosphate 5-kinase type 1 alpha
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001135636.1 |
511 |
Missense Mutation |
GCG,GTG |
A20V |
NP_001129108.1 |
NM_001135637.1 |
511 |
Missense Mutation |
GCG,GTG |
A20V |
NP_001129109.1 |
NM_001135638.1 |
511 |
Missense Mutation |
GCG,GTG |
A20V |
NP_001129110.1 |
NM_003557.2 |
511 |
Missense Mutation |
GCG,GTG |
A20V |
NP_003548.1 |
XM_005245525.4 |
511 |
Missense Mutation |
GCG,GTG |
A20V |
XP_005245582.1 |
XM_005245527.3 |
511 |
Missense Mutation |
GCG,GTG |
A20V |
XP_005245584.1 |
XM_005245528.4 |
511 |
Missense Mutation |
GCG,GTG |
A20V |
XP_005245585.1 |
XM_005245530.4 |
511 |
Intron |
|
|
XP_005245587.1 |
XM_005245531.3 |
511 |
Missense Mutation |
GCG,GTG |
A20V |
XP_005245588.1 |
XM_005245532.3 |
511 |
Intron |
|
|
XP_005245589.1 |
XM_006711563.3 |
511 |
Missense Mutation |
GCG,GTG |
A20V |
XP_006711626.1 |
XM_006711564.3 |
511 |
Missense Mutation |
GCG,GTG |
A20V |
XP_006711627.1 |
XM_006711567.3 |
511 |
Intron |
|
|
XP_006711630.1 |
XM_006711568.3 |
511 |
Intron |
|
|
XP_006711631.2 |
XM_011510043.2 |
511 |
Intron |
|
|
XP_011508345.1 |
XM_011510045.2 |
511 |
Intron |
|
|
XP_011508347.1 |
XM_011510046.2 |
511 |
UTR 5 |
|
|
XP_011508348.1 |
XM_011510048.2 |
511 |
Intron |
|
|
XP_011508350.1 |
XM_011510049.2 |
511 |
Intron |
|
|
XP_011508351.1 |
XM_017002439.1 |
511 |
Intron |
|
|
XP_016857928.1 |
XM_017002440.1 |
511 |
Intron |
|
|
XP_016857929.1 |
XM_017002441.1 |
511 |
Intron |
|
|
XP_016857930.1 |
- Gene
- VPS72
- Gene Name
- vacuolar protein sorting 72 homolog
There are no transcripts associated with this gene.
View Full Product Details