Product Details

SNP ID
rs143743518
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:158419967 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTTCTTCTCACAATTTTACAAAG[A/T]GCTGATTTGAACTCTTTATTTCTCA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
OR10K2 PubMed Links

Gene Details

Gene
OR10K2
Gene Name
olfactory receptor family 10 subfamily K member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004476.1 900 Silent Mutation GCA,GCT A300A NP_001004476.1

View Full Product Details