Product Details

SNP ID
rs143779356
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:85252657 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTTTCTTCTTTGCTGGTTCTCCT[A/G]GCTCTACTTTGCGCTTTTTAGAAGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1orf52 PubMed Links

Gene Details

Gene
C1orf52
Gene Name
chromosome 1 open reading frame 52
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198077.3 560 Missense Mutation CCA,CTA P174L NP_932343.1

View Full Product Details