Product Details

SNP ID
rs144024740
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:152623047 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGAGGCAGACACTGTGCTGGGCT[C/T]TTTGCAGGGCACTTGGGCGGAGGCT
Phenotype
MIM: 612613 MIM: 612614
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LCE3A PubMed Links

Gene Details

Gene
LCE3A
Gene Name
late cornified envelope 3A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178431.1 57 Silent Mutation AAA,AAG K19K NP_848518.1
Gene
LCE3B
Gene Name
late cornified envelope 3B
There are no transcripts associated with this gene.

View Full Product Details