Product Details

SNP ID
rs144172232
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:212856806 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCTCCATCATAGCCCGTCTCAGT[C/G]GCTTCATTCAAGTCAGCGGGGTGAG
Phenotype
MIM: 609144 MIM: 610864
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FLVCR1 PubMed Links

Gene Details

Gene
FLVCR1
Gene Name
feline leukemia virus subgroup C cellular receptor 1
There are no transcripts associated with this gene.

Gene
FLVCR1-AS1
Gene Name
FLVCR1 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

Gene
SPATA45
Gene Name
spermatogenesis associated 45
There are no transcripts associated with this gene.

View Full Product Details