Product Details

SNP ID
rs144284281
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:200873751 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACGGCCGCCGCACGTGGGTCGGGC[C/T]CGGAGGAACTCGCTGCGCATCATCT
Phenotype
MIM: 602174
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPR25 PubMed Links

Gene Details

Gene
GPR25
Gene Name
G protein-coupled receptor 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005298.3 797 Silent Mutation GCC,GCT A238A NP_005289.2

View Full Product Details