Product Details

SNP ID
rs144409534
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:145961364 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCCGTACCCTTGGCCTTGCAGGC[T/G]TCCAGCAGTGTGGCAAGGGTCTCGC
Phenotype
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
ANKRD34A PubMed Links

Gene Details

Gene
ANKRD34A
Gene Name
ankyrin repeat domain 34A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039888.3 1691 Silent Mutation GAC,GAT D132D NP_001034977.1
Gene
LIX1L
Gene Name
limb and CNS expressed 1 like
There are no transcripts associated with this gene.

Gene
POLR3GL
Gene Name
RNA polymerase III subunit G like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005277431.4 1691 Intron XP_005277488.1
XM_011510059.2 1691 Intron XP_011508361.1

View Full Product Details