Product Details

SNP ID
rs144483053
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:248180933 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAAGTCTGAGAGTGAGTTACCTCA[A/T]AAACTTTATGTTTTGCTGTTTGCTA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
OR2M2 PubMed Links

Gene Details

Gene
OR2M2
Gene Name
olfactory receptor family 2 subfamily M member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004688.1 948 Missense Mutation CAA,CAT Q316H NP_001004688.1

View Full Product Details