Product Details

SNP ID
rs144519020
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:152511721 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCTCTGGGGGCTGCTGCAGCTCT[A/G]AGGGTGGTGGCTGCTGCCTGAGCCA
Phenotype
MIM: 612619
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CRCT1 PubMed Links

Gene Details

Gene
CRCT1
Gene Name
cysteine rich C-terminal 1
There are no transcripts associated with this gene.

Gene
LCE5A
Gene Name
late cornified envelope 5A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178438.4 363 Missense Mutation AAG,GAG K63E NP_848525.1

View Full Product Details